ENA250698652 Enamine

2-(but-3-en-1-yloxy)propanoic acid

  • MDL number MFCD10689873
  • Cas Number : 566189-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56964
Availability details for
Quantity
  • Availability
  • Add to Cart