ENA250698653 Enamine

5-(4-methylphenyl)-1,2,4-triazin-3-amine

  • MDL number MFCD06739741
  • Cas Number : 886497-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56965
Availability details for
Quantity
  • Availability
  • Add to Cart