ENA250698662 Enamine

4-[(3,4-difluorophenyl)amino]-3-nitrobenzoic acid

  • MDL number MFCD02112883
  • Cas Number : 387882-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56976
Availability details for
Quantity
  • Availability
  • Add to Cart