ENA250698663 Enamine

N-hydroxyhexanamide

  • MDL number MFCD00019946
  • Cas Number : 4312-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56977
Availability details for
Quantity
  • Availability
  • Add to Cart