ENA250698664 Enamine

2-[(2-chloro-4-fluorophenoxy)methyl]-8-methylimidazo[1,2-a]pyridine

  • MDL number MFCD07345991
  • Cas Number : 871218-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56978
Availability details for
Quantity
  • Availability
  • Add to Cart