ENA250698666 Enamine

1-(2-hydroxyethyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD09865472
  • Cas Number : 64330-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56988
Availability details for
Quantity
  • Availability
  • Add to Cart