ENA250698667 Enamine

5-(2-chloroethyl)-3-(4-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD11617991
  • Cas Number : 933945-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26216
Availability details for
Quantity
  • Availability
  • Add to Cart