ENA250698671 Enamine

3-bromo-4-[(3-methyl-1,2-oxazol-5-yl)methoxy]benzoic acid

  • MDL number MFCD14705653
  • Cas Number : 1221724-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56995
Availability details for
Quantity
  • Availability
  • Add to Cart