ENA250698673 Enamine

5-bromo-2-(2-ethoxyethoxy)benzaldehyde

  • MDL number MFCD12173549
  • Cas Number : 1156314-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56997
Availability details for
Quantity
  • Availability
  • Add to Cart