ENA250698674 Enamine

3-(3-hydroxy-4-methoxyphenyl)propanoic acid

  • MDL number MFCD01444519
  • Cas Number : 1135-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56998
Availability details for
Quantity
  • Availability
  • Add to Cart