ENA250698680 Enamine

(2-aminoethyl)(ethyl)methylamine

  • MDL number MFCD09753411
  • Cas Number : 70111-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57007

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-57007
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart