ENA250698683 Enamine

3-methoxypropanamide

  • MDL number MFCD11646082
  • Cas Number : 15438-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57072
Availability details for
Quantity
  • Availability
  • Add to Cart