ENA250698690 Enamine

1-(cyclohexylmethyl)piperidin-4-amine

  • MDL number MFCD04118096
  • Cas Number : 64306-77-0
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57084
Availability details for
Quantity
  • Availability
  • Add to Cart