ENA250698695 Enamine

2-chloro-N-[5-(dimethylsulfamoyl)-2-methoxyphenyl]propanamide

  • MDL number MFCD13688062
  • Cas Number : 1155000-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27989
Availability details for
Quantity
  • Availability
  • Add to Cart