ENA250698698 Enamine

3-(2,3-dihydro-1-benzofuran-5-yl)-1-[3-(hydroxymethyl)piperidin-1-yl]prop-2-en-1-one

  • Cas Number : 1158108-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57117
Availability details for
Quantity
  • Availability
  • Add to Cart