ENA250698700 Enamine

3-{[(4-methoxyphenyl)methoxy]methyl}aniline

  • MDL number MFCD11147554
  • Cas Number : 1021042-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57119
Availability details for
Quantity
  • Availability
  • Add to Cart