ENA250698704 Enamine

3-{[4-(hydroxymethyl)piperidin-1-yl]sulfonyl}thiophene-2-carboxylic acid

  • MDL number MFCD12634800
  • Cas Number : 1097060-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57125
Availability details for
Quantity
  • Availability
  • Add to Cart