ENA250698706 Enamine

1-(4-aminobutyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD09808652
  • Cas Number : 1016685-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57127
Availability details for
Quantity
  • Availability
  • Add to Cart