ENA250698712 Enamine

2-[2-(1H-1,2,3,4-tetrazol-1-yl)acetamido]benzoic acid

  • MDL number MFCD09944470
  • Cas Number : 1016820-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57143
Availability details for
Quantity
  • Availability
  • Add to Cart