ENA250698715 Enamine

ethyl({[4-(4-methoxypiperidin-1-yl)phenyl]methyl})amine

  • MDL number MFCD11978628
  • Cas Number : 1096865-58-5
  • Molecular Weight : 248.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57147
Availability details for
Quantity
  • Availability
  • Add to Cart