ENA250698724 Enamine

5-(5-methylfuran-2-yl)-1,2,4-triazin-3-amine

  • MDL number MFCD11101843
  • Cas Number : 1152506-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57158
Availability details for
Quantity
  • Availability
  • Add to Cart