ENA250698727 Enamine

N-(5-amino-2-methylphenyl)-2-(piperidin-1-yl)acetamide

  • MDL number MFCD03117507
  • Cas Number : 459803-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57172
Availability details for
Quantity
  • Availability
  • Add to Cart