ENA250698728 Enamine

3-phenyl-2-[(pyridin-3-yl)formamido]propanoic acid

  • MDL number MFCD00420584
  • Cas Number : 36724-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57174
Availability details for
Quantity
  • Availability
  • Add to Cart