ENA250698733 Enamine

7-hydroxy-3,4-dihydrospiro[1-benzopyran-2,1'-cyclohexane]-4-one

  • MDL number MFCD08741814
  • Cas Number : 62756-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57180

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57180
Availability details for
Quantity
  • Availability
  • Add to Cart