ENA250698734 Enamine

4-chloro-3-{[(4-sulfamoylphenyl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD06367429
  • Cas Number : 794573-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16092
Availability details for
Quantity
  • Availability
  • Add to Cart