ENA250698740 Enamine

3-[(thiophen-2-ylsulfanyl)methyl]-1-benzofuran-2-carboxylic acid

  • MDL number MFCD12469026
  • Cas Number : 1039891-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57189
Availability details for
Quantity
  • Availability
  • Add to Cart