ENA250698741 Enamine

4-(aminomethyl)-N-cyclohexyl-N-methylaniline

  • MDL number MFCD07365255
  • Cas Number : 893753-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57190
Availability details for
Quantity
  • Availability
  • Add to Cart