ENA250698750 Enamine

4-(1H-1,3-benzodiazol-1-ylmethyl)-N'-hydroxybenzene-1-carboximidamide

  • MDL number MFCD09928964
  • Cas Number : 1016739-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237...239
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57224
Availability details for
Quantity
  • Availability
  • Add to Cart