ENA250698751 Enamine

5-chloro-2-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD08443046
  • Cas Number : 21792-87-0
  • Molecular Weight : 256.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-64
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29724

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-29724
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart