ENA250698754 Enamine

6-(1H-imidazol-1-yl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD14705677
  • Cas Number : 110714-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239-241
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-57231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57231
Availability details for
Quantity
  • Availability
  • Add to Cart