ENA250698758 Enamine

6-phenyl-4,5,6,7-tetrahydro-1,3-benzothiazol-2-amine

  • MDL number MFCD01366561
  • Cas Number : 79932-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57235
Availability details for
Quantity
  • Availability
  • Add to Cart