ENA250698762 Enamine

1-(3,4-dimethoxyphenyl)-2-(2-methylphenyl)ethan-1-amine

  • MDL number MFCD09810563
  • Cas Number : 1017346-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57242
Availability details for
Quantity
  • Availability
  • Add to Cart