ENA250698765 Enamine

1-(4-fluorophenyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD11202249
  • Cas Number : 1038333-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57245
Availability details for
Quantity
  • Availability
  • Add to Cart