ENA250698767 Enamine

5-(4-methanesulfonylphenyl)-1,2,4-triazin-3-amine

  • MDL number MFCD11134086
  • Cas Number : 1094231-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57247
Availability details for
Quantity
  • Availability
  • Add to Cart