ENA250698774 Enamine

N-(2,2-dichloroethenyl)propanamide

  • MDL number MFCD00222318
  • Cas Number : 37001-11-9
  • Molecular Weight : 168.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-57260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57260
Availability details for
Quantity
  • Availability
  • Add to Cart