ENA250698776 Enamine

1-(3-aminophenyl)-3-ethylurea

  • MDL number MFCD11115880
  • Cas Number : 41402-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57277
Availability details for
Quantity
  • Availability
  • Add to Cart