ENA250698780 Enamine

4-(1,2,3,4-tetrahydroquinoline-1-carbonyl)aniline

  • MDL number MFCD07359508
  • Cas Number : 137975-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57285
Availability details for
Quantity
  • Availability
  • Add to Cart