ENA250698781 Enamine

4-[cyclohexyl(methyl)amino]benzoic acid

  • MDL number MFCD11156540
  • Cas Number : 243662-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57288
Availability details for
Quantity
  • Availability
  • Add to Cart