ENA250698786 Enamine

4-(2H-1,3-benzodioxol-5-yl)butan-2-amine

  • MDL number MFCD09732717
  • Cas Number : 40742-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33168
Availability details for
Quantity
  • Availability
  • Add to Cart