ENA250698787 Enamine

1-(1-methyl-1H-1,3-benzodiazol-2-yl)ethan-1-ol

  • MDL number MFCD01050195
  • Cas Number : 3319-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57294
Availability details for
Quantity
  • Availability
  • Add to Cart