ENA250698791 Enamine

3-{[(prop-2-en-1-yl)amino]methyl}benzonitrile

  • MDL number MFCD11138034
  • Cas Number : 1019561-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-57298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57298
Availability details for
Quantity
  • Availability
  • Add to Cart