ENA250698798 Enamine

4-cyanobutane-1-sulfonyl chloride

  • MDL number MFCD14684119
  • Cas Number : 35517-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57308
Availability details for
Quantity
  • Availability
  • Add to Cart