ENA250698815 Enamine

4-[butyl(methyl)amino]benzoic acid

  • MDL number MFCD11156535
  • Cas Number : 75681-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58544
Availability details for
Quantity
  • Availability
  • Add to Cart