ENA250698817 Enamine

4-(pyridin-3-yloxy)benzonitrile

  • MDL number MFCD11178092
  • Cas Number : 58835-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58576
Availability details for
Quantity
  • Availability
  • Add to Cart