ENA250698842 Enamine

6-(2,6-difluorobenzoyl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD13661392
  • Cas Number : 1097107-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58859
Availability details for
Quantity
  • Availability
  • Add to Cart