ENA250698846 Enamine

N'-hydroxy-2-(piperidin-1-ylmethyl)benzene-1-carboximidamide

  • MDL number MFCD09935230
  • Cas Number : 1016780-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58867

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58867
Availability details for
Quantity
  • Availability
  • Add to Cart