ENA250698850 Enamine

6-(4-fluorophenoxy)-N'-hydroxypyridine-3-carboximidamide

  • MDL number MFCD09951166
  • Cas Number : 1017043-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58871
Availability details for
Quantity
  • Availability
  • Add to Cart