ENA250698855 Enamine

N-cyclohexylpiperidine-4-carboxamide hydrochloride

  • MDL number MFCD08456915
  • Cas Number : 1019851-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223...225
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58878

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58878
Availability details for
Quantity
  • Availability
  • Add to Cart