ENA250698859 Enamine

2-(chloromethyl)-1H-benzimidazole hydrochloride

  • MDL number MFCD00159934
  • Cas Number : 91862-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35245

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-35245
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart