ENA250698861 Enamine

3-[4-(chlorosulfonyl)-2-methylphenoxy]propanoic acid

  • MDL number MFCD09944876
  • Cas Number : 1016822-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58884
Availability details for
Quantity
  • Availability
  • Add to Cart